Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000489.4 | NP_000480.3 | 2 | 324 | ||
NM_138270.3 | NP_612114.2 | 2 | 324 | |||
XM_005262153.4 | XP_005262210.2 | 2 | 324 | |||
XM_005262154.4 | XP_005262211.2 | 2 | 324 | |||
XM_005262155.4 | XP_005262212.2 | 2 | 425 | |||
XM_005262157.4 | XP_005262214.2 | 2 | 324 | |||
XM_006724666.3 | XP_006724729.1 | 2 | 324 | |||
XM_006724668.2 | XP_006724731.1 | 2 | 324 | |||
XM_017029601.1 | XP_016885090.1 | 2 | 324 | |||
XM_017029602.1 | XP_016885091.1 | 2 | 408 | |||
XM_017029604.1 | XP_016885093.1 | 2 | 324 | |||
XM_017029605.1 | XP_016885094.1 | 2 | 425 | |||
XM_017029606.1 | XP_016885095.1 | 2 | 425 | |||
XM_017029607.1 | XP_016885096.1 | 2 | 425 | |||
XM_017029609.1 | XP_016885098.1 | 2 | 425 | |||
XM_017029610.1 | XP_016885099.1 | 2 | 425 | |||
GenBank mRNA | AB102641.1 | - | 2 | 56 | ||
AB208928.1 | - | 2 | 255 | |||
AK308419.1 | - | 2 | 263 | |||
BC002521.2 | - | 2 | 146 | |||
BT007188.1 | - | 1 | 56 | |||
DA759777.1 | - | 2 | 425 | |||
DA791316.1 | - | 2 | 324 | |||
U72936.2 | - | 2 | 271 | |||
U72937.2 | - | 2 | 271 | |||
U72938.2 | - | 2 | 271 |