Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111893.2 | 24 | 3924 |
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 24 | 4064 | ||
GenBank mRNA | AF333769.2 | - | 24 | 3932 | ||
BC132737.1 | - | 23 | 3726 | 40146759 | ||
BC144130.1 | - | 22 | 3634 |
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201380.2 | NP_001188309.2 | 24 | 4235 | ||
GenBank mRNA | BX538190.1 | - | 5 | 499 |