siRNA Details
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135821.1 | NP_001129293.1 | 3, 4 | 511 | ||
NM_001135822.1 | NP_001129294.1 | 2, 3 | 368 | |||
NM_001242824.1 | NP_001229753.1 | 2, 3 | 389 | |||
NM_002004.3 | NP_001995.1 | 3, 4 | 566 | |||
XM_005244962.1 | XP_005245019.1 | 2, 3 | 279 | |||
XM_005244963.1 | XP_005245020.1 | 2, 3 | 334 | |||
GenBank mRNA | AK021828.1 | - | 1, 2 | 1302 | ||
AK022841.1 | - | 1, 2 | 1277 | |||
AK291084.1 | - | 3, 4 | 367 | |||
BC010004.2 | - | 2, 3 | 355 | 4132071 | ||
BP370037.1 | - | 2, 3 | 393 | |||
CN346026.1 | - | 2, 3 | 338 | |||
D14697.1 | - | 3, 4 | 461 | |||
DA610405.1 | - | 2, 3 | 334 | |||
J05262.1 | - | 1, 2 | 155 | |||
M29863.1 | - | 1, 2 | 129 |
Gene Symbol: FDPSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003262.1 | 1 | 3995 | |||
GenBank mRNA | BC050292.1 | - | 2 | 2719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
