Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195728.2 | NP_001182657.1 | 12 | 4000 | ||
NM_001252652.1 | NP_001239581.1 | 12 | 4065 | |||
NM_004171.3 | NP_004162.2 | 11 | 4362 | |||
XM_005253067.1 | XP_005253124.1 | 11 | 3839 | |||
XM_011520285.2 | XP_011518587.1 | 11 | 3876 | |||
XM_017018136.1 | XP_016873625.1 | 11 | 4659 | |||
XM_017018137.1 | XP_016873626.1 | 11 | 3879 | |||
XM_017018138.1 | XP_016873627.1 | 11 | 3863 | |||
XM_017018139.1 | XP_016873628.1 | 10 | 3904 | |||
GenBank mRNA | AY066021.1 | - | 11 | 4049 | ||
BC132768.1 | - | 11 | 3929 | 40146790 | ||
CR627022.1 | - | 1 | 371 |
Gene Symbol: ADCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516909.1 | XP_011515211.1 | 1 | 59 | ||
GenBank mRNA | HY037845.1 | - | 2 | 157 |
Gene Symbol: AIFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130846.3 | NP_001124318.2 | 1 | 70 | ||
GenBank mRNA | DQ016496.2 | - | 1 | 70 |
Gene Symbol: ALDH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000692.4 | NP_000683.3 | 2 | 2828 | ||
XM_011517802.2 | XP_011516104.1 | 2 | 2768 | |||
GenBank mRNA | BC001619.1 | - | 2 | 2764 | 3356389 |
Gene Symbol: ATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290048.1 | NP_001276977.1 | 13 | 4949 | ||
NM_015459.4 | NP_056274.3 | 13 | 5051 | |||
XM_006718493.1 | XP_006718556.1 | 12 | 4876 | |||
GenBank mRNA | AK090822.1 | - | 1 | 135 | ||
BX648468.1 | - | 1 | 1564 | |||
CR936784.1 | - | 13 | 4921 |
Gene Symbol: ATP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208828.1 | - | 7 | 3394 |
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 5 | 2745 | ||
NM_001243767.1 | NP_001230696.1 | 5 | 2610 | |||
NM_152265.4 | NP_689478.1 | 6 | 2812 |
Gene Symbol: C11orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271680.4 | XP_005271737.1 | 9 | 4721 | ||
XM_011543009.2 | XP_011541311.1 | 9 | 4929 |
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543218.2 | XP_011541520.1 | 6 | 1308 | ||
GenBank mRNA | BC037547.1 | - | 5 | 1229 |
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093805.1 | - | 1 | 546 | ||
AK097957.1 | - | 1 | 107 |
Gene Symbol: CPAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527920.2 | XP_011526222.1 | 42 | 5892 | ||
XM_011527924.2 | XP_011526226.1 | 42 | 5818 | |||
GenBank mRNA | AK124052.1 | - | 2 | 1118 |
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001876.3 | NP_001867.2 | 19 | 4115 | ||
XM_005273762.2 | XP_005273819.1 | 19 | 4091 | |||
GenBank mRNA | AJ420378.1 | - | 1 | 1710 |
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020432.1 | XP_016875921.1 | 1 | 83 |
Gene Symbol: ENOSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127219.1 | - | 1 | 760 |
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 11 | 2917 | ||
NM_001284343.1 | NP_001271272.1 | 7 | 1995 | |||
NM_007270.4 | NP_009201.2 | 10 | 2758 | |||
GenBank mRNA | AB209352.1 | - | 3 | 4478 | ||
AK075331.1 | - | 10 | 2721 | |||
AK128597.1 | - | 11 | 2700 | |||
AL050187.1 | - | 2 | 1086 | |||
BC007443.2 | - | 4 | 1475 | |||
BC064418.1 | - | 5 | 1681 | |||
BC072422.1 | - | 4 | 1424 | |||
BC099918.1 | - | 3 | 1320 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 6 | 1735 | |||
NR_027339.1 | 4 | 1839 | ||||
NR_027340.1 | 3 | 1468 | ||||
NR_027342.1 | 4 | 1749 | ||||
GenBank mRNA | BC011872.2 | - | 4 | 1750 |
Gene Symbol: FLJ33534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311163.1 | - | 2 | 588 |
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000148.3 | NP_000139.1 | 4 | 2734 | ||
XM_006723127.2 | XP_006723190.1 | 3 | 2726 | |||
XM_017026552.1 | XP_016882041.1 | 3 | 2646 | |||
XM_017026553.1 | XP_016882042.1 | 4 | 2560 | |||
GenBank mRNA | M35531.1 | - | 2 | 1862 |
Gene Symbol: GFOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514699.2 | XP_011513001.1 | 2 | 3481 |
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128816.1 | - | 1 | 172 |
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537563.1 | - | 9 | 3194 |
Gene Symbol: HS3ST3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006041.2 | NP_006032.1 | 2 | 2356 | ||
GenBank mRNA | BC063301.1 | - | 2 | 2266 | 30343352 |
Gene Symbol: IQCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB324323.1 | - | 1 | 175 |
Gene Symbol: LINC00508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126452.2 | 4 | 399 | |||
GenBank mRNA | BC032019.1 | - | 1 | 88 | ||
DA807329.1 | - | 4 | 401 |
Gene Symbol: LINC01252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033890.1 | 3 | 543 | |||
GenBank mRNA | BC041827.1 | - | 3 | 546 |
Gene Symbol: LOC105369325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ999767.1 | - | 2 | 1161 |
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209235.1 | - | 1036 |
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127174.1 | NP_001120646.1 | 6 | 4159 | ||
NM_001127175.1 | NP_001120647.1 | 6 | 4270 | |||
NM_001127176.1 | NP_001120648.1 | 7 | 4426 | |||
NM_031939.3 | NP_114145.2 | 8 | 4529 | |||
XM_005258365.3 | XP_005258422.1 | 7 | 4339 | |||
XM_011526226.2 | XP_011524528.1 | 7 | 4310 | |||
XM_017026035.1 | XP_016881524.1 | 7 | 4306 | |||
XM_017026036.1 | XP_016881525.1 | 7 | 4310 | |||
XM_017026037.1 | XP_016881526.1 | 6 | 4299 |
Gene Symbol: MRS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY333753.1 | - | 3413 |
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167738.1 | NP_001161210.1 | 27 | 10479 | ||
NM_020443.4 | NP_065176.3 | 30 | 11819 | |||
XM_006711609.2 | XP_006711672.2 | 28 | 10454 | |||
XM_006711610.2 | XP_006711673.2 | 27 | 10430 | |||
XM_006711611.1 | XP_006711674.1 | 28 | 10570 | |||
XM_011510097.1 | XP_011508399.1 | 29 | 11489 | |||
XM_011510098.1 | XP_011508400.1 | 29 | 11474 | |||
XM_011510099.1 | XP_011508401.1 | 28 | 11465 | |||
XM_011510100.1 | XP_011508402.1 | 29 | 11327 | |||
XM_011510101.1 | XP_011508403.1 | 28 | 11303 | |||
XM_011510102.1 | XP_011508404.1 | 27 | 11294 | |||
XM_017002751.1 | XP_016858240.1 | 28 | 11318 | |||
XM_017002752.1 | XP_016858241.1 | 26 | 10421 | |||
GenBank mRNA | AK091567.1 | - | 1 | 424 |
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011538419.2 | XP_011536721.1 | 12 | 5721 | ||
XM_011538420.2 | XP_011536722.1 | 11 | 5388 | |||
XM_017019364.1 | XP_016874853.1 | 13 | 6034 | |||
XM_017019365.1 | XP_016874854.1 | 12 | 5762 | |||
XM_017019366.1 | XP_016874855.1 | 12 | 5749 | |||
XM_017019367.1 | XP_016874856.1 | 11 | 5587 |
Gene Symbol: PANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028078.1 | XP_016883567.1 | 1 | 769 |
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU361470.1 | - | 1 | 88 | ||
GU361471.1 | - | 1, 2 | 88 | |||
KC357594.1 | - | 1, 2 | 88 | |||
KC357595.1 | - | 3 | 185 | |||
KC774171.1 | - | 1 | 88 |
Gene Symbol: PAX8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047570.1 | 3 | 255 | |||
GenBank mRNA | AK056052.1 | - | 3 | 255 | ||
AK126431.1 | - | 1 | 1583 | |||
AK130275.1 | - | 3 | 234 | |||
AK310158.1 | - | 6 | 693 | |||
DA573927.1 | - | 3 | 245 |
Gene Symbol: PQBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ973601.1 | - | 3 | 263 | ||
AJ973602.1 | - | 2 | 480 | |||
AJ973603.1 | - | 3 | 263 |
Gene Symbol: PRKAR2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109996.1 | 2 | 927 | |||
NR_109997.1 | 3 | 848 | ||||
GenBank mRNA | AK127190.1 | - | 2 | 906 |
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004283.3 | NP_004274.1 | 5 | 3298 | ||
GenBank mRNA | AF019226.1 | - | 1 | 2321 | ||
BC035309.1 | - | 1 | 244 |
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092794.1 | - | 1 | 474 |
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005253317.4 | XP_005253374.1 | 1 | 55 | ||
XM_017018843.1 | XP_016874332.1 | 1 | 55 | |||
XM_017018845.1 | XP_016874334.1 | 1 | 55 |
Gene Symbol: SYT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB037848.1 | - | 6 | 2967 | ||
AL512743.1 | - | 6 | 2904 |
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146213.1 | NP_001139685.2 | 17 | 3360 | ||
NM_001146214.1 | NP_001139686.1 | 18 | 3452 | |||
NM_022771.4 | NP_073608.4 | 18 | 3411 | |||
NR_027449.1 | 16 | 3149 |
Gene Symbol: TCEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094425.1 | - | 761 |
Gene Symbol: TPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152263.3 | NP_689476.2 | 10 | 3621 |
Gene Symbol: ZDHHC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536661.2 | XP_011534963.1 | 1 | 1834 |
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 1 | 54 | ||
NM_001278287.1 | NP_001265216.1 | 1 | 54 | |||
GenBank mRNA | AK309719.1 | - | 1 | 242 |
Gene Symbol: ZNF660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005265090.4 | XP_005265147.1 | 2 | 4813 | ||
XM_006713117.3 | XP_006713180.1 | 3 | 4814 | |||
XM_011533622.2 | XP_011531924.1 | 3 | 5018 | |||
GenBank mRNA | BX538171.1 | - | 1 | 4135 |
Gene Symbol: ZNF829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936826.1 | - | 5 | 1614 |