Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025396.1 | XP_016880885.1 | 2 | 655 | ||
XM_017025397.1 | XP_016880886.1 | 2 | 655 | |||
XM_017025399.1 | XP_016880888.1 | 1 | 13 | |||
XM_017025400.1 | XP_016880889.1 | 2 | 655 | |||
XM_017025401.1 | XP_016880890.1 | 2 | 655 | |||
XM_017025404.1 | XP_016880893.1 | 2 | 631 | |||
XM_017025405.1 | XP_016880894.1 | 1 | 291 | |||
XM_017025410.1 | XP_016880899.1 | 2 | 636 | |||
XM_017025416.1 | XP_016880905.1 | 2 | 651 |
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523857.2 | XP_011522159.1 | 5 | 4537 |
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014683.3 | NP_055498.3 | 27 | 7082 | ||
GenBank mRNA | AL080134.1 | - | 1 | 1039 |