siRNA Details
Gene Symbol: TRIM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006355.4 | NP_006346.1 | 8 | 2978 | ||
XM_005248799.4 | XP_005248856.1 | 8 | 2707 | |||
XM_005248800.4 | XP_005248857.1 | 8 | 2848 | |||
GenBank mRNA | AB209906.1 | - | 6 | 5268 | ||
CR936707.1 | - | 4 | 6186 | |||
U90547.1 | - | 6 | 2410 |
Gene Symbol: CAPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097277.1 | - | 1 | 100 |
Gene Symbol: CEP57L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715462.3 | XP_006715525.1 | 12 | 6434 | ||
XM_011535762.2 | XP_011534064.1 | 9 | 5658 | |||
XM_017010768.1 | XP_016866257.1 | 11 | 6198 | |||
XM_017010769.1 | XP_016866258.1 | 11 | 6086 | |||
XM_017010771.1 | XP_016866260.1 | 9 | 5663 |
Gene Symbol: DISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522125.2 | XP_011520427.1 | 10 | 9516 | ||
XM_017022687.1 | XP_016878176.1 | 11 | 9265 |
Gene Symbol: DNAJC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304944.1 | NP_001291873.1 | 4 | 4176 | ||
NM_024902.3 | NP_079178.2 | 3 | 3714 |
Gene Symbol: F2RL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003950.2 | NP_003941.2 | 2 | 1600 | ||
GenBank mRNA | AF055917.1 | - | 2 | 1600 | ||
AF080214.1 | - | 2 | 1427 |
Gene Symbol: KIAA1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526114.1 | XP_011524416.1 | 26 | 3562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
