siRNA Details
Gene Symbol: DDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160147.1 | NP_001153619.1 | 13 | 11758 | ||
NM_001160148.1 | NP_001153620.1 | 13 | 11821 | |||
NM_030637.2 | NP_085140.2 | 12 | 11737 | |||
XM_011537188.2 | XP_011535490.1 | 15 | 11933 | |||
GenBank mRNA | AK124546.1 | - | 1 | 2158 |
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053561.1 | - | 5 | 1267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
