siRNA Details
Gene Symbol: APOA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000482.3 | NP_000473.2 | 3 | 1420 | ||
GenBank mRNA | M10373.1 | - | 1 | 914 | ||
M13654.1 | - | 3 | 1363 | |||
M14566.1 | - | 2 | 1252 | |||
X13629.1 | - | 3 | 1357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
