Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204817.1 | NP_001191746.1 | 3 | 2108 | ||
NM_032828.3 | NP_116217.1 | 3 | 2111 | |||
GenBank mRNA | AK160374.1 | - | 2 | 1987 | ||
BC126237.1 | - | 3 | 1973 | 8991953 | ||
BC126239.1 | - | 3 | 1973 | 8991955 |
Gene Symbol: GINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021067.4 | NP_066545.3 | 7 | 1806 | ||
NR_134574.1 | 8 | 1928 | ||||
XM_017028162.1 | XP_016883651.1 | 7 | 1763 | |||
GenBank mRNA | D80008.1 | - | 7 | 1761 |
Gene Symbol: HR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833253.1 | - | 1 | 1635 |
Gene Symbol: LOC285638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833206.1 | - | 3 | 3920 |
Gene Symbol: ZCWPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK299989.1 | - | 5 | 957 |