siRNA Details
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 15 | 3725 | ||
NR_033717.1 | 16 | 2666 | ||||
XM_006724458.1 | XP_006724521.1 | 14 | 3537 | |||
XM_017029221.1 | XP_016884710.1 | 14 | 3607 | |||
GenBank mRNA | AK022824.1 | - | 3 | 1973 | ||
AL833937.1 | - | 4 | 951 | |||
BC051913.1 | - | 16 | 2608 | |||
BC064617.1 | - | 15 | 3682 | 5575894 | ||
Y18004.1 | - | 15 | 3659 |
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163942.1 | NP_001157414.1 | 6 | 1775 | ||
GenBank mRNA | BX452500.1 | - | 1 | 767 |
Gene Symbol: ARHGAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130670.1 | - | 1 | 393 |
Gene Symbol: ARHGAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258328.2 | NP_001245257.1 | 23 | 4947 | ||
NM_001282334.1 | NP_001269263.1 | 13 | 4834 | |||
NM_001282335.1 | NP_001269264.1 | 22 | 4513 | |||
NM_001321232.1 | NP_001308161.1 | 23 | 4883 | |||
NM_012292.4 | NP_036424.2 | 23 | 5036 |
Gene Symbol: ATP6V1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001690.3 | NP_001681.2 | 15 | 2815 | ||
GenBank mRNA | AF113129.1 | - | 15 | 2745 | ||
AF339827.1 | - | 3 | 1177 | |||
BC013138.1 | - | 15 | 2763 | 4454722 |
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028034.1 | XP_016883523.1 | 3 | 446 |
Gene Symbol: C10orf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153244.1 | NP_694976.1 | 2 | 1006 | ||
GenBank mRNA | BC029034.1 | - | 2 | 1006 | 5194035 | |
BC036401.1 | - | 1 | 23 |
Gene Symbol: CARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032587.3 | NP_115976.2 | 3 | 3571 | ||
XM_017009989.1 | XP_016865478.1 | 2 | 3124 | |||
XM_017009990.1 | XP_016865479.1 | 4 | 3805 | |||
GenBank mRNA | AF356193.1 | - | 3 | 3695 |
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208814.1 | - | 12 | 4653 |
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090536.1 | - | 1 | 73 |
Gene Symbol: CEP57L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266935.4 | XP_005266992.3 | 11 | 5210 | ||
XM_005266936.4 | XP_005266993.3 | 11 | 5210 | |||
XM_005266937.4 | XP_005266994.3 | 11 | 5162 | |||
XM_005266938.4 | XP_005266995.3 | 11 | 5159 | |||
XM_006715462.3 | XP_006715525.1 | 12 | 5574 | |||
XM_011535759.2 | XP_011534061.1 | 11 | 5524 | |||
XM_011535762.2 | XP_011534064.1 | 9 | 4798 | |||
XM_017010764.1 | XP_016866253.1 | 12 | 5713 | |||
XM_017010765.1 | XP_016866254.1 | 13 | 5968 | |||
XM_017010767.1 | XP_016866256.1 | 11 | 5524 | |||
XM_017010768.1 | XP_016866257.1 | 11 | 5338 | |||
XM_017010769.1 | XP_016866258.1 | 11 | 5226 | |||
XM_017010770.1 | XP_016866259.1 | 13 | 5917 | |||
XM_017010771.1 | XP_016866260.1 | 9 | 4803 | |||
XM_017010772.1 | XP_016866261.1 | 10 | 5262 |
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714037.3 | XP_006714100.1 | 6 | 7032 |
Gene Symbol: CHMP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020412.4 | NP_065145.2 | 1 | 2674 |
Gene Symbol: CREM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209533.1 | - | 1 | 1264 |
Gene Symbol: DECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055839.1 | - | 4 | 361 | ||
AK225961.1 | - | 3 | 280 |
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 1807 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 1904 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 1785 | |||
NM_004402.3 | NP_004393.1 | 7 | 1735 | |||
NR_104222.1 | 8 | 1872 | ||||
NR_135150.1 | 8 | 1859 | ||||
NR_135151.1 | 8 | 1822 | ||||
NR_135152.1 | 7 | 1757 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 1670 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 1686 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 1441 | |||
GenBank mRNA | AB028911.1 | - | 8 | 1652 | ||
AB028912.1 | - | 7 | 1698 | |||
AB028913.1 | - | 8 | 1616 | |||
AF064019.1 | - | 7 | 1529 | |||
AK027141.1 | - | 1477 | ||||
BC032827.1 | - | 8 | 1684 | |||
BC048797.1 | - | 7 | 1721 | 5204996 |
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027858.1 | - | 1 | 1327 |
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008019.1 | XP_016863508.1 | 2 | 486 |
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145064.2 | NP_001138536.1 | 9 | 6041 | ||
GenBank mRNA | AK123110.1 | - | 1 | 2887 | ||
BC048422.1 | - | 1 | 342 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027409.1 | 9 | 1118 | |||
XM_017022018.1 | XP_016877507.1 | 7 | 899 | |||
GenBank mRNA | AK123221.1 | - | 5 | 463 | ||
BX648160.1 | - | 8 | 1073 | |||
BX648575.1 | - | 8 | 825 |
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027410.1 | 9 | 1104 | |||
XM_017022210.1 | XP_016877699.1 | 8 | 1096 | |||
XM_017022214.1 | XP_016877703.1 | 8 | 1096 | |||
XM_017022215.1 | XP_016877704.1 | 8 | 1096 | |||
XM_017022216.1 | XP_016877705.1 | 8 | 1125 | |||
XM_017022217.1 | XP_016877706.1 | 7 | 1047 | |||
XM_017022218.1 | XP_016877707.1 | 8 | 1723 | |||
GenBank mRNA | BX647673.1 | - | 8 | 1073 |
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128816.1 | - | 2 | 1775 |
Gene Symbol: HKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026699.1 | XP_016882188.1 | 1 | 2928 |
Gene Symbol: IMPAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017813.4 | NP_060283.3 | 5 | 3542 | ||
GenBank mRNA | AK090988.1 | - | 1 | 295 |
Gene Symbol: IRGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007561.2 | NP_001007562.1 | 3 | 6945 | ||
XM_005258515.3 | XP_005258572.1 | 3 | 7024 | |||
GenBank mRNA | AK022583.1 | - | 1 | 1024 | ||
AK096168.1 | - | 1 | 1737 | |||
AK124786.1 | - | 1 | 2451 | |||
AK128554.1 | - | 1 | 2579 |
Gene Symbol: JPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK056172.1 | - | 3 | 397 | ||
AV714079.1 | - | LIT1011 Jpx |
400 | |||
BP871721.1 | - | 3 | 365 |
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054769.1 | - | 1 | 1091 |
Gene Symbol: LINC01089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094482.1 | - | 1 | 59 | ||
AK095700.1 | - | 1 | 1009 |
Gene Symbol: LOC100130278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094114.1 | - | 1 | 2152 |
Gene Symbol: LOC101927954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121565.1 | 3 | 3017 | |||
GenBank mRNA | AK126072.1 | - | 3 | 3018 |
Gene Symbol: LOC339803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023856.1 | - | 2 | 2228 |
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005278385.4 | XP_005278442.1 | 6 | 3913 | ||
GenBank mRNA | BC034279.1 | - | 1 | 2062 |
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028105.1 | XP_016883594.1 | 19 | 8888 | ||
XM_017028107.1 | XP_016883596.1 | 12 | 8978 |
Gene Symbol: MIR34AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132742.1 | 2 | 3153 | |||
GenBank mRNA | EF570048.1 | - | 2 | 2920 |
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058101.1 | - | 1 | 681 |
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008528.2 | NP_001008528.1 | 4 | 1190 | ||
XM_005257382.2 | XP_005257439.1 | 3 | 1276 | |||
XM_011524830.1 | XP_011523132.1 | 4 | 1076 | |||
GenBank mRNA | AK123486.1 | - | 1 | 1198 |
Gene Symbol: NABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI038029.1 | - | 1 | 344 |
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 12 | 4206 | |||
GenBank mRNA | AB209623.1 | - | 12 | 3994 |
Gene Symbol: PLEKHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022835.2 | NP_073746.2 | 19 | 6760 | ||
XM_005259163.2 | XP_005259220.1 | 19 | 6581 | |||
XM_006723334.2 | XP_006723397.1 | 19 | 7111 | |||
XM_011527232.2 | XP_011525534.1 | 19 | 6687 | |||
XM_017027150.1 | XP_016882639.1 | 18 | 7252 | |||
XM_017027151.1 | XP_016882640.1 | 18 | 7243 |
Gene Symbol: PLOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124035.1 | - | 1 | 949 |
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282736.1 | NP_001269665.1 | 13 | 9223 | ||
NM_032812.8 | NP_116201.7 | 14 | 9370 |
Gene Symbol: POGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005244999.2 | XP_005245056.1 | 1 | 1808 | ||
XM_017000745.1 | XP_016856234.1 | 1 | 1809 |
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK301029.1 | - | 5 | 1059 |
Gene Symbol: RBM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516370.2 | XP_011514672.1 | 20 | 3887 | ||
XM_017012389.1 | XP_016867878.1 | 18 | 3663 | |||
XM_017012390.1 | XP_016867879.1 | 17 | 3555 |
Gene Symbol: RNF139-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108047.1 | 2 | 2123 |
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127914.1 | - | 1 | 1278 |
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270440.1 | NP_001257369.1 | 7 | 1082 | ||
GenBank mRNA | AK093672.1 | - | 7 | 1082 |
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA445450.1 | - | 1 | 239 |
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936667.1 | - | 6 | 1332 |
Gene Symbol: SNTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080537.1 | NP_001074006.1 | 4 | 1363 |
Gene Symbol: STX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052874.4 | NP_443106.1 | 10 | 2282 | ||
XM_017022893.1 | XP_016878382.1 | 10 | 2037 | |||
GenBank mRNA | BC038359.1 | - | 7 | 1805 |
Gene Symbol: TLCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164407.1 | NP_001157879.1 | 4 | 4687 |
Gene Symbol: TRPV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018727.5 | NP_061197.4 | 16 | 3223 | ||
NM_080704.3 | NP_542435.2 | 17 | 3340 | |||
NM_080705.3 | NP_542436.2 | 16 | 3266 | |||
NM_080706.3 | NP_542437.2 | 15 | 3592 | |||
GenBank mRNA | AF196175.1 | - | 16 | 3278 | ||
AF196176.1 | - | 16 | 3264 | |||
AL136801.1 | - | 17 | 3336 | |||
BC132820.1 | - | 15 | 3544 | 40146842 | ||
BC136633.1 | - | 15 | 3544 | 9020623 |
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055832.1 | - | 2 | 1255 |
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000551.3 | NP_000542.1 | 3 | 2008 | ||
NM_198156.2 | NP_937799.1 | 2 | 1885 | |||
GenBank mRNA | AA361862.1 | - | 1 | 74 | ||
BG284971.1 | - | 1 | 318 | |||
BG288777.1 | - | 1 | 195 |
Gene Symbol: ZNF205-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024166.1 | 3 | 941 | |||
NR_024167.1 | 3 | 936 | ||||
GenBank mRNA | BC001809.2 | - | 3 | 941 | ||
BC001824.2 | - | 3 | 941 | |||
BC020202.1 | - | 2 | 424 | |||
BC121099.1 | - | 3 | 563 | |||
BC121100.1 | - | 3 | 568 |
Gene Symbol: ZNF260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012756.2 | NP_001012774.1 | 4 | 4857 | ||
NM_001166036.1 | NP_001159508.1 | 4 | 4851 | |||
NM_001166037.1 | NP_001159509.1 | 3 | 4743 | |||
NM_001166038.1 | NP_001159510.1 | 3 | 4724 | |||
XM_011526897.2 | XP_011525199.1 | 2 | 4666 | |||
XM_017026741.1 | XP_016882230.1 | 2 | 5171 | |||
GenBank mRNA | AK123830.1 | - | 2 | 3250 | ||
BC042676.1 | - | 4 | 4779 | 4821139 |
Gene Symbol: ZNF536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128073.1 | - | 1 | 537 |
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 5 | 10566 |
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258903.4 | XP_005258960.1 | 5 | 5029 | ||
XM_011526950.2 | XP_011525252.1 | 5 | 5110 |
Gene Symbol: ZNF843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096248.1 | - | 3 | 1510 |
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 9 | 5528 | ||
NM_032173.3 | NP_115549.2 | 9 | 5321 | |||
XM_011530435.2 | XP_011528737.1 | 10 | 5517 | |||
XM_011530436.2 | XP_011528738.1 | 11 | 5572 | |||
XM_011530437.1 | XP_011528739.1 | 10 | 6242 | |||
XM_011530438.2 | XP_011528740.1 | 10 | 5758 | |||
XM_017028990.1 | XP_016884479.1 | 10 | 6013 | |||
GenBank mRNA | AB051436.1 | - | 9 | 5394 | ||
AK022809.1 | - | 1 | 2144 | |||
BC042614.1 | - | 1 | 2357 | |||
BC051344.1 | - | 1 | 2449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -

















