siRNA Details
Gene Symbol: APOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000039.2 | NP_000030.1 | 4 | 1115 | ||
NM_001318017.1 | NP_001304946.1 | 4 | 828 | |||
NM_001318018.1 | NP_001304947.1 | 4 | 820 | |||
NM_001318021.1 | NP_001304950.1 | 4 | 1071 | |||
GenBank mRNA | A14829.1 | - | 3 | 768 | ||
A15879.1 | - | 2 | 677 | |||
AK292231.1 | - | 4 | 789 | |||
AL531353.3 | - | 4 | 820 | |||
AL557908.3 | - | 4 | 829 | |||
BC005380.1 | - | 4 | 787 | 3934992 | ||
BC110286.1 | - | 4 | 810 | 5164525 | ||
BU600916.1 | - | 4 | 747 | |||
M11791.1 | - | 3 | 749 | |||
M27875.1 | - | 3 | 768 | |||
M29068.1 | - | 2 | 695 | |||
X00566.1 | - | 3 | 784 | |||
X02162.1 | - | 4 | 835 |
Gene Symbol: ZFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI207587.1 | - | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
