Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 7551 | ||
NM_001308340.1 | NP_001295269.1 | 3 | 3799 | |||
XM_005268665.4 | XP_005268722.1 | 3 | 7332 | |||
GenBank mRNA | AK123102.1 | - | 1 | 380 |
Gene Symbol: APLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647107.1 | - | 1 | 967 |
Gene Symbol: CHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018223.2 | NP_060693.2 | 6 | 546 | ||
GenBank mRNA | AK001658.1 | - | 6 | 526 |
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022059.3 | NP_071342.2 | 6 | 1431 | ||
GenBank mRNA | AF275260.1 | - | 6 | 942 | ||
AF301016.1 | - | 6 | 1322 | |||
AF337812.1 | - | 6 | 856 | |||
AK027389.1 | - | 6 | 1349 | |||
BC044930.1 | - | 6 | 1126 | 5762458 | ||
JC209418.1 | - | 6 | 1431 |
Gene Symbol: FLJ10038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026891.1 | 1 | 3530 |
Gene Symbol: GK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039547.2 | NP_001034636.1 | 16 | 3594 | ||
NR_033289.1 | 17 | 4677 | ||||
XM_011512624.2 | XP_011510926.1 | 16 | 3392 | |||
XM_017006089.1 | XP_016861578.1 | 17 | 3657 | |||
XM_017006090.1 | XP_016861579.1 | 15 | 3505 | |||
GenBank mRNA | BC032470.1 | - | 18 | 2850 | 5217372 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124130.1 | - | 1 | 1540 |
Gene Symbol: ILDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199351.2 | NP_955383.1 | 10 | 7440 | ||
XM_006711314.3 | XP_006711377.1 | 5 | 8299 | |||
XM_017001252.1 | XP_016856741.1 | 10 | 7612 | |||
XM_017001253.1 | XP_016856742.1 | 10 | 7747 | |||
XM_017001254.1 | XP_016856743.1 | 9 | 7555 | |||
XM_017001255.1 | XP_016856744.1 | 9 | 7435 | |||
XM_017001256.1 | XP_016856745.1 | 7 | 7231 |
Gene Symbol: LOC730179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126076.1 | - | 2 | 2047 |
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126313.1 | - | 1 | 398 |
Gene Symbol: NKAPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537594.1 | - | 1 | 4458 |
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU617706.1 | - | 1 | 648 |
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022353.2 | NP_071748.2 | 9 | 1866 | ||
XM_005246766.3 | XP_005246823.1 | 9 | 1629 | |||
XM_006712685.1 | XP_006712748.1 | 9 | 1741 | |||
XM_006712686.1 | XP_006712749.1 | 9 | 1612 | |||
XM_011511631.1 | XP_011509933.1 | 9 | 1760 | |||
XM_017004676.1 | XP_016860165.1 | 9 | 2379 | |||
XM_017004677.1 | XP_016860166.1 | 9 | 1633 | |||
XM_017004678.1 | XP_016860167.1 | 9 | 1578 | |||
XM_017004679.1 | XP_016860168.1 | 9 | 1597 | |||
XM_017004680.1 | XP_016860169.1 | 9 | 1719 | |||
XM_017004681.1 | XP_016860170.1 | 9 | 1584 | |||
XM_017004682.1 | XP_016860171.1 | 9 | 2362 | |||
XM_017004683.1 | XP_016860172.1 | 9 | 1449 | |||
XM_017004684.1 | XP_016860173.1 | 9 | 1521 | |||
GenBank mRNA | AK055441.1 | - | 9 | 1766 | ||
BC011904.2 | - | 9 | 1435 | 4121450 |
Gene Symbol: PDCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209040.1 | - | 2 | 1461 |
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002725.3 | NP_002716.1 | 3 | 4676 | ||
NM_201348.1 | NP_958505.1 | 3 | 4666 |
Gene Symbol: PSMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199779.1 | NP_001186708.1 | 6 | 4010 | ||
NM_001199780.1 | NP_001186709.1 | 5 | 3854 | |||
NM_002794.4 | NP_002785.1 | 6 | 4378 |
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF461898.1 | - | 1 | 47 |
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090801.1 | - | 1 | 85 |
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832047.1 | - | 2 | 1170 |
Gene Symbol: SLC36A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537583.2 | XP_011535885.1 | 1 | 1403 | ||
XM_011537586.2 | XP_011535888.1 | 1 | 337 |
Gene Symbol: SPACA6P-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108100.1 | 2 | 2289 | |||
GenBank mRNA | AK125996.1 | - | 2 | 2288 |
Gene Symbol: TMEM231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM973146.1 | - | 1 | 148 |
Gene Symbol: TMEM9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073431.1 | 5 | 967 | |||
GenBank mRNA | AA339546.1 | - | 3 | 278 |