Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009909.3 | NP_001009909.2 | 12 | 3921 | ||
NM_001252008.1 | NP_001238937.1 | 12 | 3917 | |||
NM_001252010.1 | NP_001238939.1 | 10 | 3795 | |||
GenBank mRNA | AK126974.1 | - | 4 | 2964 | ||
AL832641.3 | - | 12 | 3832 | |||
BC042863.1 | - | 1 | 481 |
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 12 | 4033 | ||
NM_017457.5 | NP_059431.1 | 13 | 4036 | |||
GenBank mRNA | BC036615.1 | - | 1 | 1904 | ||
BC047631.1 | - | 1 | 1899 |
Gene Symbol: IRGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007561.2 | NP_001007562.1 | 3 | 2419 | ||
XM_005258515.3 | XP_005258572.1 | 3 | 2498 |
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204171.1 | NP_001191100.1 | 10 | 9439 | ||
NM_001204172.1 | NP_001191101.1 | 3 | 8611 | |||
NM_001278516.1 | NP_001265445.1 | 10 | 9521 | |||
NM_001278517.1 | NP_001265446.1 | 8 | 9295 | |||
NM_001278518.1 | NP_001265447.1 | 7 | 9110 | |||
NM_001278519.1 | NP_001265448.1 | 5 | 8920 | |||
NM_002393.4 | NP_002384.2 | 11 | 9589 |
Gene Symbol: MRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252091.1 | NP_001239020.1 | 1 | 176 | ||
NM_012219.4 | NP_036351.3 | 1 | 176 | |||
GenBank mRNA | AK124954.1 | - | 1 | 127 | ||
DA235195.1 | - | 1 | 176 | |||
DA355883.1 | - | 1 | 59 |
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016279.1 | XP_016871768.1 | 5 | 1734 | ||
GenBank mRNA | AB208817.1 | - | 5 | 1724 |
Gene Symbol: PRKAR2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109996.1 | 2 | 2249 | |||
NR_109997.1 | 3 | 2170 | ||||
GenBank mRNA | AK127190.1 | - | 2 | 2228 |
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085400.1 | NP_001078869.1 | 7 | 1829 | ||
XM_017008590.1 | XP_016864079.1 | 5 | 1497 | |||
GenBank mRNA | AK025431.1 | - | 2 | 590 |
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI711397.1 | - | 1 | 267 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 2 | 256 | |||
GenBank mRNA | AK289466.1 | - | 3 | 225 |
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128061.1 | - | 1 | 379 |
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537742.1 | - | 1 | 1525 |