Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002124.3 | NP_002115.2 | 1 | 127 | ||
GenBank mRNA | AK289463.1 | - | 1 | 96 | ||
AK291987.1 | - | 1 | 127 | |||
AY961072.1 | - | 1 | 33 | |||
AY961073.1 | - | 1 | 33 | |||
BC033827.1 | - | 1 | 76 | 5216243 | ||
BC108922.1 | - | 1 | 88 | 40023604 | ||
DQ002917.1 | - | 1 | 33 | |||
HG974574.1 | - | 1 | 33 | |||
HG974596.1 | - | 1 | 33 | |||
HG974600.1 | - | 1 | 33 | |||
HM067861.1 | - | 1 | 33 | |||
HM067862.1 | - | 1 | 33 | |||
HM067863.1 | - | 1 | 33 | |||
KJ534866.1 | - | 1 | 33 | |||
L02545.1 | - | 1 | 47 | |||
M11161.1 | - | 1 | 33 | |||
M20430.1 | - | 1 | 76 | |||
M20504.1 | - | 1 | 68 | |||
M28583.1 | - | 1 | 72 | |||
M30179.1 | - | 1 | 33 | |||
M33600.1 | - | 1 | 103 | |||
X03069.1 | - | 1 | 95 |