Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021141.3 | NP_066964.1 | 21 | 2476 | ||
GenBank mRNA | AK096408.1 | - | 18 | 2878 | ||
AK222603.1 | - | 21 | 2393 | |||
AK290740.1 | - | 21 | 2362 | |||
BC019027.2 | - | 21 | 2344 | 4555162 | ||
BC095442.1 | - | 21 | 2362 | 30329657 | ||
J04977.1 | - | 21 | 2369 | |||
M30938.1 | - | 21 | 2363 |