Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137669.1 | NP_001131141.1 | 7 | 1492 | ||
XM_005245137.1 | XP_005245194.1 | 8 | 1582 | |||
XM_011509494.2 | XP_011507796.1 | 8 | 1582 | |||
XM_011509496.1 | XP_011507798.1 | 8 | 1582 | |||
XM_011509500.1 | XP_011507802.1 | 8 | 1582 | |||
XM_011509503.1 | XP_011507805.1 | 8 | 1582 | |||
XM_017001188.1 | XP_016856677.1 | 8 | 1582 | |||
XM_017001189.1 | XP_016856678.1 | 8 | 1613 | |||
XM_017001190.1 | XP_016856679.1 | 8 | 2393 | |||
XM_017001191.1 | XP_016856680.1 | 8 | 1594 | |||
XM_017001192.1 | XP_016856681.1 | 8 | 1883 | |||
XM_017001193.1 | XP_016856682.1 | 8 | 1710 | |||
XM_017001194.1 | XP_016856683.1 | 9 | 1730 | |||
XM_017001195.1 | XP_016856684.1 | 5 | 466 | |||
XM_017001196.1 | XP_016856685.1 | 2 | 88 | |||
XM_017001197.1 | XP_016856686.1 | 5 | 466 | |||
GenBank mRNA | AF510428.1 | - | 1 | 18 | ||
AK302768.1 | - | 8 | 1572 | |||
AL136902.1 | - | 7 | 1561 | |||
BC121032.2 | - | 1 | 18 | |||
BC121033.1 | - | 1 | 18 | |||
BC142944.1 | - | 7 | 1467 | 8860326 |