Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080821.2 | NP_543011.2 | 3 | 1187 | ||
GenBank mRNA | AJ311123.1 | - | 3 | 1148 | ||
AK075565.1 | - | 3 | 1134 | |||
BC017725.1 | - | 3 | 1144 | 3887324 |