Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080821.2 | NP_543011.2 | 3 | 1508 | ||
GenBank mRNA | AJ311123.1 | - | 3 | 1469 | ||
AK075565.1 | - | 3 | 1455 | |||
AL133555.1 | - | 1 | 191 | |||
BC017725.1 | - | 3 | 1465 | 3887324 | ||
L17329.1 | - | 1 | 232 |