Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077399.2 | NP_001070867.1 | 1 | 99 | ||
NM_015488.4 | NP_056303.3 | 1 | 99 | |||
GenBank mRNA | AF318057.1 | - | 1 | 99 | ||
AF417001.1 | - | 1 | 99 |