siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003097.4 | NP_003088.1 | 3 | 403 | ||
NM_022805.3 | NP_073716.1 | 5 | 598 | |||
NM_022806.3 | NP_073717.1 | 5 | 608 | |||
NM_022807.3 | NP_073718.1 | 6 | 743 | |||
NM_022808.3 | NP_073719.1 | 5 | 635 | |||
GenBank mRNA | AF319522.1 | - | 3 | 262 | ||
AK222633.1 | - | 5 | 558 | |||
BC003180.1 | - | 3 | 309 | 2958068 | ||
BC018809.2 | - | 3 | 298 | |||
BC024777.1 | - | 4 | 431 | 3907606 | ||
BC025178.1 | - | 3 | 275 | 4158469 | ||
DA131564.1 | - | 5 | 552 | |||
DA369087.1 | - | 3 | 404 | |||
J04615.1 | - | 3 | 308 | |||
L80004.1 | - | 5 | 587 | |||
L80005.1 | - | 6 | 733 | |||
U41303.1 | - | 3 | 308 | |||
X15892.1 | - | 1 | 185 |
Gene Symbol: SNURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005678.4 | NP_005669.2 | 3 | 403 | ||
NM_022804.2 | NP_073715.1 | 3 | 373 | |||
GenBank mRNA | AF101044.1 | - | 3 | 308 | ||
AK098563.1 | - | 3 | 307 | |||
DA134019.1 | - | 3 | 373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




