siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003097.4 | NP_003088.1 | 2, 3 | 272 | ||
NM_022805.3 | NP_073716.1 | 4, 5 | 467 | |||
NM_022806.3 | NP_073717.1 | 4, 5 | 477 | |||
NM_022807.3 | NP_073718.1 | 5, 6 | 612 | |||
NM_022808.3 | NP_073719.1 | 4, 5 | 504 | |||
GenBank mRNA | AF319522.1 | - | 2, 3 | 131 | ||
AK222633.1 | - | 4, 5 | 427 | |||
BC003180.1 | - | 2, 3 | 178 | 2958068 | ||
BC018809.2 | - | 2, 3 | 167 | |||
BC024777.1 | - | 3, 4 | 300 | 3907606 | ||
BC025178.1 | - | 2, 3 | 144 | 4158469 | ||
DA069028.1 | - | 4, 5 | 477 | |||
DA131564.1 | - | 4, 5 | 421 | |||
DA369087.1 | - | 2, 3 | 273 | |||
J04615.1 | - | 2, 3 | 177 | |||
L80004.1 | - | 4, 5 | 456 | |||
L80005.1 | - | 5, 6 | 602 | |||
U41303.1 | - | 2, 3 | 177 |
Gene Symbol: SNURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005678.4 | NP_005669.2 | 2, 3 | 272 | ||
NM_022804.2 | NP_073715.1 | 2, 3 | 242 | |||
GenBank mRNA | AF101044.1 | - | 2, 3 | 177 | ||
AK098563.1 | - | 2, 3 | 176 | |||
DA134019.1 | - | 2, 3 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




