Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 9 | 1563 | ||
XM_005247950.3 | XP_005248007.1 | 9 | 1560 | |||
XM_011513406.2 | XP_011511708.1 | 9 | 1592 | |||
XM_011513407.2 | XP_011511709.1 | 9 | 1592 | |||
XM_011513408.1 | XP_011511710.1 | 10 | 1704 | |||
XM_011513409.2 | XP_011511711.1 | 10 | 1675 | |||
GenBank mRNA | AK093044.1 | - | 5 | 569 | ||
BC035655.1 | - | 9 | 1063 | 5588410 |