siRNA Details
Gene Symbol: SH3PXD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017995.2 | NP_001017995.1 | 5 | 548 | ||
NM_001308175.1 | NP_001295104.1 | 5 | 548 | |||
XM_017009351.1 | XP_016864840.1 | 5 | 548 | |||
GenBank mRNA | AB430862.1 | - | 5 | 377 | ||
AK128871.1 | - | 5 | 548 | |||
BC038561.1 | - | 5 | 383 | |||
DQ109556.1 | - | 5 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
