Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300981.1 | NP_001287910.1 | 5 | 553 | ||
NM_021130.4 | NP_066953.1 | 4 | 396 | |||
GenBank mRNA | AB451307.1 | - | 4 | 307 | ||
AB451438.1 | - | 4 | 307 | |||
AK026569.1 | - | 1 | 351 | |||
AK130101.1 | - | 4 | 352 | |||
AK290085.1 | - | 4 | 352 | |||
AK290851.1 | - | 4 | 354 | |||
AK293003.1 | - | 5 | 509 | |||
AK316564.1 | - | 4 | 351 | |||
BC000689.2 | - | 1 | 329 | 3349335 | ||
BC003026.1 | - | 1 | 335 | 2823490 | ||
BC005320.1 | - | 1 | 351 | 3935025 | ||
BC005982.1 | - | 1 | 354 | 4109260 | ||
BC013915.1 | - | 1 | 356 | 3684721 | ||
BC018843.2 | - | 1 | 386 | |||
BC073992.1 | - | 1 | 341 | 6161389 | ||
BC093076.1 | - | 5 | 418 | 30414589 | ||
BC106030.1 | - | 1 | 343 | 2986147 | ||
BC137057.1 | - | 4 | 396 | 9021054 | ||
BC137058.1 | - | 4 | 396 | 9021055 | ||
BM806224.1 | - | 5 | 407 | |||
BM909285.1 | - | 73 | ||||
BU786475.1 | - | 4 | 390 | |||
CD014106.1 | - | 5 | 509 | |||
CR456707.1 | - | 4 | 307 | |||
EU794650.1 | - | 1 | 343 | |||
Y00052.1 | - | 4 | 321 |
Gene Symbol: PPIAL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143883.2 | NP_001137355.1 | 1 | 382 | ||
GenBank mRNA | AB084917.1 | - | 1 | 382 |
Gene Symbol: PPIAL4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135789.3 | NP_001129261.2 | 1 | 382 | ||
GenBank mRNA | BC130376.1 | - | 1 | 340 | 40146407 | |
BC130378.1 | - | 1 | 340 | 40146409 |
Gene Symbol: PPIAL4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164261.1 | NP_001157733.1 | 1 | 382 |
Gene Symbol: PPIAL4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144032.2 | NP_001137504.2 | 1 | 382 |
Gene Symbol: PPIAL4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164262.2 | NP_001157734.2 | 1 | 382 |
Gene Symbol: PPIAL4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123068.1 | NP_001116540.1 | 1 | 340 |
Gene Symbol: PPIAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036506.1 | 1 | 361 | |||
GenBank mRNA | BC157831.1 | - | 1 | 361 | 9057050 | |
BC171823.1 | - | 1 | 361 | 9054477 |