Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 18 | 2361 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 18 | 2349 | |||
GenBank mRNA | AK292534.1 | - | 18 | 2232 | ||
BC110417.1 | - | 18 | 2320 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 18 | 2349 | |||
GenBank mRNA | BC063309.1 | - | 19 | 2323 | ||
DQ309037.1 | - | 18 | 2323 |
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 18 | 2349 | |||
GenBank mRNA | AK057241.1 | - | 16 | 1827 | ||
AK302625.1 | - | 16 | 1774 | |||
AL833871.1 | - | 7 | 829 | |||
BC144672.1 | - | 15 | 1954 | |||
BC150302.1 | - | 18 | 2323 | 8860399 | ||
DQ309038.1 | - | 16 | 1797 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 18 | 2361 | |||
GenBank mRNA | BC066774.1 | - | 18 | 2335 | ||
BC099913.1 | - | 17 | 3176 | 4838463 | ||
BC125193.1 | - | 17 | 2141 | 40122746 | ||
BC125194.1 | - | 17 | 2141 | 40122749 |