siRNA Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 4 | 427 | ||
NR_111943.1 | 3 | 633 | ||||
NR_111944.1 | 4 | 427 | ||||
GenBank mRNA | AK311951.1 | - | 4 | 335 | ||
BC009407.2 | - | 1 | 318 | 4334552 | ||
BC019899.1 | - | 1 | 323 | 3882924 | ||
BC020453.1 | - | 1 | 322 | |||
BC022370.1 | - | 4 | 340 | 4738331 | ||
BC049824.1 | - | 1 | 315 | 6212543 | ||
BC062715.1 | - | 4 | 425 | 4068842 | ||
BC071928.1 | - | 1 | 326 | 5090053 | ||
BE905890.1 | - | 3 | 303 | |||
BI597659.1 | - | 3 | 633 | |||
BM017772.1 | - | 4 | 429 | |||
M13932.1 | - | 4 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
