siRNA Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 1 | 20 | ||
NR_111944.1 | 1 | 20 | ||||
GenBank mRNA | BC062715.1 | - | 1 | 18 | 4068842 | |
BM017772.1 | - | 1 | 21 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
