Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000184.2 | NP_000175.1 | 2 | 330 | ||
GenBank mRNA | AF110493.1 | - | 2 | 224 | ||
AK290492.1 | - | 2 | 330 | |||
BC010914.1 | - | 2 | 334 | 4134713 | ||
BC029387.1 | - | 2 | 360 | 4762402 | ||
BC130457.1 | - | 2 | 325 | 40146488 | ||
BC130459.1 | - | 2 | 325 | 40146490 | ||
M15386.1 | - | 2 | 294 | |||
X55656.1 | - | 2 | 257 |
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 2 | 330 | ||
GenBank mRNA | BC010913.1 | - | 2 | 336 | 4133117 | |
BC020719.1 | - | 2 | 330 | 4732536 |