siRNA Details
Gene Symbol: HBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000559.2 | NP_000550.2 | 1 | 79 | ||
GenBank mRNA | BC010913.1 | - | 1 | 85 | 4133117 | |
BC020719.1 | - | 1 | 79 | 4732536 |
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000184.2 | NP_000175.1 | 1 | 79 | ||
GenBank mRNA | AK290492.1 | - | 1 | 79 | ||
BC010914.1 | - | 1 | 83 | 4134713 | ||
BC029387.1 | - | 1 | 109 | 4762402 | ||
BC130457.1 | - | 1 | 74 | 40146488 | ||
BC130459.1 | - | 1 | 74 | 40146490 | ||
M15386.1 | - | 1 | 43 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
