Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 2 | 318 | ||
NR_037648.1 | 2 | 337 | ||||
XM_011542895.2 | XP_011541197.1 | 2 | 415 | |||
XM_011542896.2 | XP_011541198.1 | 2 | 204 | |||
XM_017018000.1 | XP_016873489.1 | 2 | 293 | |||
XM_017018001.1 | XP_016873490.1 | 2 | 417 | |||
XM_017018003.1 | XP_016873492.1 | 2 | 293 | |||
XM_017018004.1 | XP_016873493.1 | 2 | 415 | |||
XM_017018005.1 | XP_016873494.1 | 1 | 1450 | |||
XM_017018006.1 | XP_016873495.1 | 2 | 293 | |||
GenBank mRNA | AF103801.1 | - | 2 | 216 | ||
AF447877.1 | - | 1 | 565 | |||
AK023987.1 | - | 2 | 187 | |||
AK295267.1 | - | 2 | 273 | |||
AK295428.1 | - | 2 | 187 | |||
AL136923.1 | - | 2 | 198 | |||
BC013902.2 | - | 2 | 174 | 3956972 | ||
DA056004.1 | - | 2 | 318 |