siRNA Details
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 1 | 129 | ||
NM_145660.2 | NP_663693.1 | 1 | 129 | |||
GenBank mRNA | AY014914.1 | - | 1 | 130 | ||
AY014915.1 | - | 1 | 129 | |||
BC006276.2 | - | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
