siRNA Details
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 1, 2 | 205 | ||
NM_145660.2 | NP_663693.1 | 1, 2 | 205 | |||
GenBank mRNA | AK290466.1 | - | 1, 2 | 86 | ||
AK311506.1 | - | 1, 2 | 178 | |||
AK313637.1 | - | 1, 2 | 142 | |||
AY014914.1 | - | 1, 2 | 206 | |||
AY014915.1 | - | 1, 2 | 205 | |||
BC006276.2 | - | 1, 2 | 115 | |||
DA865632.1 | - | 1, 2 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
