siRNA Details
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 2 | 229 | ||
NM_145660.2 | NP_663693.1 | 2 | 229 | |||
GenBank mRNA | AK290466.1 | - | 2 | 110 | ||
AK313637.1 | - | 2 | 166 | |||
AY014914.1 | - | 2 | 230 | |||
AY014915.1 | - | 2 | 229 | |||
BC006276.2 | - | 2 | 139 |
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136540.1 | NP_001130012.1 | 2 | 211 | ||
NM_001136541.1 | NP_001130013.1 | 2 | 172 | |||
NM_003661.3 | NP_003652.2 | 2 | 172 | |||
NM_145343.2 | NP_663318.1 | 3 | 326 | |||
XM_005261796.3 | XP_005261853.1 | 2 | 151 | |||
XM_011530478.2 | XP_011528780.1 | 2 | 110 | |||
GenBank mRNA | AF019225.2 | - | 2 | 171 | ||
AF305224.1 | - | 2 | 111 | |||
AF305428.1 | - | 2 | 151 | |||
AF323540.1 | - | 3 | 325 | |||
AK296099.1 | - | 2 | 121 | |||
AK300454.1 | - | 2 | 141 | |||
AK303919.1 | - | 2 | 149 | |||
AK309143.1 | - | 2 | 169 | |||
AK313752.1 | - | 3 | 265 | |||
BC017331.1 | - | 2 | 102 | |||
BC112943.1 | - | 2 | 127 | 4512933 | ||
BC113867.1 | - | 2 | 102 | |||
BC114475.1 | - | 2 | 102 | |||
BC127186.1 | - | 2 | 102 | 40034503 | ||
BC141823.1 | - | 2 | 127 | 40148156 | ||
BC142720.1 | - | 2 | 127 | |||
BC143038.1 | - | 2 | 102 | 40034505 | ||
BC143039.1 | - | 2 | 102 | 40034506 | ||
BG419709.1 | - | 1 | 41 | |||
DA375861.1 | - | 3 | 267 | |||
DC340792.1 | - | 2 | 149 | |||
DC367266.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
