siRNA Details
Gene Symbol: PHLDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007350.3 | NP_031376.3 | 2 | 3689 | ||
GenBank mRNA | BC047362.1 | - | 1 | 226 |
Gene Symbol: ANO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142679.1 | NP_001136151.1 | 20 | 3535 | ||
GenBank mRNA | AK310641.1 | - | 20 | 3502 |
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721447.2 | XP_006721510.2 | 1 | 803 | ||
XM_017024182.1 | XP_016879671.1 | 1 | 803 |
Gene Symbol: LOC101927751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135827.1 | 2 | 1188 | |||
GenBank mRNA | AK126286.1 | - | 2 | 1188 |
Gene Symbol: LPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167671.2 | NP_001161143.1 | 11 | 7688 | ||
NM_001167672.2 | NP_001161144.1 | 10 | 7216 | |||
NM_005578.4 | NP_005569.1 | 11 | 7745 | |||
XM_005247446.4 | XP_005247503.1 | 11 | 7722 | |||
XM_005247450.4 | XP_005247507.1 | 10 | 7691 | |||
XM_005247451.4 | XP_005247508.1 | 11 | 7722 | |||
XM_005247453.2 | XP_005247510.1 | 10 | 7694 | |||
XM_011512820.2 | XP_011511122.1 | 13 | 7989 | |||
XM_011512823.2 | XP_011511125.1 | 10 | 9804 | |||
XM_011512827.2 | XP_011511129.1 | 11 | 7645 | |||
XM_011512828.2 | XP_011511130.1 | 11 | 7810 | |||
XM_011512831.2 | XP_011511133.1 | 10 | 7552 | |||
XM_011512833.2 | XP_011511135.1 | 12 | 7779 | |||
XM_011512834.2 | XP_011511136.1 | 12 | 7870 | |||
XM_017006377.1 | XP_016861866.1 | 13 | 7851 | |||
XM_017006378.1 | XP_016861867.1 | 12 | 7867 | |||
XM_017006379.1 | XP_016861868.1 | 12 | 8002 | |||
XM_017006380.1 | XP_016861869.1 | 12 | 7898 | |||
XM_017006382.1 | XP_016861871.1 | 9 | 7569 |
Gene Symbol: POLD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209274.1 | - | 1 | 1129 |
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015012.3 | NP_055827.1 | 7 | 1784 | ||
GenBank mRNA | BC035034.1 | - | 7 | 1662 | 5259179 | |
BC044597.1 | - | 7 | 1652 | 4811565 | ||
BC064366.1 | - | 7 | 1539 | |||
D26067.1 | - | 7 | 1588 |
Gene Symbol: ZSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK003500.1 | - | 1 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
