Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000558.4 | NP_000549.1 | 2 | 196 | ||
GenBank mRNA | AF097635.1 | - | 2 | 153 | ||
AF105974.1 | - | 2 | 151 | |||
AF147332.1 | - | 2 | 148 | |||
AF349571.1 | - | 2 | 155 | |||
AK223392.1 | - | 2 | 173 | |||
BC005931.1 | - | 2 | 167 | 4047053 | ||
BC008572.1 | - | 2 | 153 | 4177243 | ||
BC032122.2 | - | 2 | 153 | 4849981 | ||
BC050661.1 | - | 2 | 153 | 5209215 | ||
BC101846.1 | - | 2 | 160 | 8069352 | ||
BC101848.1 | - | 2 | 160 | 8069354 | ||
V00493.1 | - | 2 | 167 |
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 2 | 196 |
Gene Symbol: HBZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005332.2 | NP_005323.1 | 2 | 185 | ||
XM_005255287.3 | XP_005255344.1 | 3 | 815 | |||
XM_005255288.3 | XP_005255345.1 | 3 | 932 | |||
GenBank mRNA | BC027892.1 | - | 2 | 165 | 5224569 | |
CR456848.1 | - | 2 | 130 | |||
M24173.1 | - | 2 | 159 |