Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1MW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048181.2 | NP_001041646.1 | 2 | 348 |
Gene Symbol: OPN1LW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020061.5 | NP_064445.2 | 2 | 326 | ||
GenBank mRNA | BQ639996.1 | - | 2 | 269 | ||
CR749814.1 | - | 2 | 284 | |||
DN693335.1 | - | 2 | 326 | |||
EL952084.1 | - | 2 | 301 |
Gene Symbol: OPN1MW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000513.2 | NP_000504.1 | 2 | 348 | ||
GenBank mRNA | BC140938.1 | - | 2 | 348 | 9056750 | |
BC143790.1 | - | 2 | 329 | 9052304 | ||
BC171754.1 | - | 2 | 329 | 9053255 | ||
BC171755.1 | - | 2 | 329 | 9053256 |
Gene Symbol: OPN1MW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003960089.4 | XP_003960138.1 | 2 | 266 |