Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097615.2 | NP_001091084.2 | 2 | 225 | ||
XM_017012372.1 | XP_016867861.1 | 2 | 264 | |||
XM_017012373.1 | XP_016867862.1 | 2 | 264 | |||
GenBank mRNA | AF468111.1 | - | 2 | 171 | ||
AJ277740.1 | - | 2 | 125 | |||
AJ277741.1 | - | 2 | 125 | |||
AK309487.1 | - | 2 | 187 | |||
BC141857.1 | - | 2 | 184 | |||
DA574442.1 | - | 2 | 188 | |||
DA580738.1 | - | 2 | 187 | |||
DB106772.1 | - | 2 | 226 |
Gene Symbol: POLR2J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006234.4 | NP_006225.1 | 2 | 202 | ||
XM_005250452.4 | XP_005250509.1 | 2 | 267 | |||
XM_017012353.1 | XP_016867842.1 | 2 | 199 | |||
GenBank mRNA | AK290515.1 | - | 2 | 199 | ||
BC024165.1 | - | 2 | 168 | 4069069 | ||
BC040116.1 | - | 2 | 163 | |||
BC065711.1 | - | 2 | 180 | 4133200 | ||
BC139902.1 | - | 2 | 165 | 40147225 | ||
BC141830.1 | - | 2 | 164 | 40148671 | ||
BM545237.1 | - | 2 | 320 | |||
L37127.1 | - | 2 | 189 | |||
X82385.1 | - | 2 | 189 | |||
X98433.1 | - | 2 | 156 |
Gene Symbol: POLR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032959.5 | NP_116581.3 | 2 | 253 | ||
GenBank mRNA | AB209398.1 | - | 2 | 191 | ||
AJ277739.1 | - | 2 | 125 | |||
BC056864.1 | - | 2 | 143 | |||
BC086857.1 | - | 2 | 253 | 6007730 |
Gene Symbol: POLR2J4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003655.3 | 2 | 209 | |||
GenBank mRNA | AK296066.1 | - | 2 | 190 | ||
BC017250.2 | - | 2 | 185 | |||
BC017341.2 | - | 2 | 173 | |||
BC062722.1 | - | 2 | 168 | |||
BC105584.1 | - | 2 | 191 | |||
BX537878.1 | - | 2 | 164 | |||
BX538054.1 | - | 2 | 166 | |||
DA147922.1 | - | 2 | 209 |