Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCA1 SIRNA Assays ›
          Gene Symbol:
          ABCA1
          Gene Name:
          ATP binding cassette subfamily A member 1
          Gene Aliases:
          ABC-1, ABC1, CERP, HDLDT1, TGD
          Chromosome Location:
          Chr.9: 104781002 - 104928246 on Build GRCh38
          UniGene ID:
          Hs.659274
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2508

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABCA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005502.3 NP_005493.2 28, 29 4448
          XM_005251773.2 XP_005251830.1 28, 29 4363
          XM_005251776.2 XP_005251833.1 27, 28 4203
          XM_011518339.2 XP_011516641.1 29, 30 4438
          XM_011518340.2 XP_011516642.1 30, 31 4653
          XM_011518341.2 XP_011516643.1 29, 30 4432
          XM_011518342.2 XP_011516644.1 26, 27 4141
          XM_011518344.2 XP_011516646.1 29, 30 4438
          XM_017014378.1 XP_016869867.1 29, 30 4636
          XM_017014379.1 XP_016869868.1 29, 30 5044
          XM_017014380.1 XP_016869869.1 29, 30 4370
          XM_017014381.1 XP_016869870.1 29, 30 4436
          XM_017014382.1 XP_016869871.1 27, 28 4901
          GenBank mRNA AB055982.1 - 27, 28 4044
          AB445477.1 - 27, 28 4044
          AF165281.1 - 26, 27 3984
          AF285167.1 - 28, 29 4334
          AJ012376.1 - 26, 27 3984
          BC146856.1 - 28, 29 4258 9056784

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          peptide secretion
          protein lipidation
          phagocytosis, engulfment
          lysosome organization
          G-protein coupled receptor signaling pathway
          response to nutrient
          cholesterol metabolic process
          negative regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          endosomal transport
          positive regulation of cAMP biosynthetic process
          intracellular cholesterol transport
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          high-density lipoprotein particle assembly
          response to laminar fluid shear stress
          apolipoprotein A-I-mediated signaling pathway
          lipoprotein metabolic process
          response to drug
          cholesterol homeostasis
          reverse cholesterol transport
          phospholipid translocation
          interleukin-1 beta secretion
          phospholipid homeostasis
          response to low-density lipoprotein particle
          platelet dense granule organization
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to cholesterol
          anion transmembrane transport
          receptor binding
          protein binding
          ATP binding
          phospholipid binding
          phospholipid transporter activity
          anion transmembrane transporter activity
          cholesterol binding
          cholesterol transporter activity
          syntaxin binding
          small GTPase binding
          apolipoprotein binding
          apolipoprotein A-I binding
          apolipoprotein A-I receptor activity
          ATPase activity, coupled to transmembrane movement of substances
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0