Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161779.1 | NP_001155251.1 | 3 | 603 | ||
NM_001161780.1 | NP_001155252.1 | 3 | 477 | |||
NM_001161781.1 | NP_001155253.1 | 2 | 464 | |||
NM_018444.3 | NP_060914.2 | 2 | 521 | |||
XM_011517135.2 | XP_011515437.1 | 3 | 374 | |||
XM_011517136.1 | XP_011515438.1 | 3 | 481 | |||
XM_017013587.1 | XP_016869076.1 | 2 | 536 | |||
XM_017013588.1 | XP_016869077.1 | 3 | 444 | |||
GenBank mRNA | AF155661.1 | - | 3 | 491 | ||
AK057079.1 | - | 2 | 433 | |||
AK126862.1 | - | 1 | 898 | |||
AK302651.1 | - | 2 | 407 | |||
BC047619.1 | - | 2 | 442 | 4824168 | ||
BC064978.1 | - | 3 | 461 | |||
BC098343.1 | - | 1 | 275 | 40010016 | ||
DA240240.1 | - | 2 | 466 | |||
DB084539.1 | - | 2 | 521 | |||
DC369809.1 | - | 2 | 478 |