Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU6F1 SIRNA Assays ›
Gene Symbol:
POU6F1
Gene Name:
POU class 6 homeobox 1
Gene Aliases:
BRN5, MPOU, TCFB1
Chromosome Location:
Chr.12: 51186936 - 51218134 on Build GRCh38
UniGene ID:
Hs.555886
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251311
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU6F1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002702.3
NP_002693.3
6
2242
NR_026893.1
11
2700
XM_017019507.1
XP_016874996.1
11
3887
XM_017019508.1
XP_016874997.1
12
3798
XM_017019509.1
XP_016874998.1
10
2807
XM_017019510.1
XP_016874999.1
11
3236
XM_017019511.1
XP_016875000.1
11
3405
XM_017019512.1
XP_016875001.1
11
2761
XM_017019513.1
XP_016875002.1
11
3884
XM_017019514.1
XP_016875003.1
11
3881
XM_017019515.1
XP_016875004.1
11
3227
XM_017019516.1
XP_016875005.1
11
3878
XM_017019517.1
XP_016875006.1
10
3758
XM_017019518.1
XP_016875007.1
10
3752
XM_017019519.1
XP_016875008.1
10
3712
XM_017019520.1
XP_016875009.1
9
3114
XM_017019521.1
XP_016875010.1
9
3133
XM_017019522.1
XP_016875011.1
10
2611
XM_017019523.1
XP_016875012.1
8
2318
XM_017019524.1
XP_016875013.1
10
2709
XM_017019525.1
XP_016875014.1
11
2697
XM_017019526.1
XP_016875015.1
10
2605
XM_017019527.1
XP_016875016.1
10
2624
XM_017019528.1
XP_016875017.1
11
3130
XM_017019529.1
XP_016875018.1
10
3016
XM_017019530.1
XP_016875019.1
9
2921
GenBank mRNA
AK095191.1
-
11
2700
AL832881.1
-
6
2242
BC051326.1
-
6
2045
6275848
DA026332.1
-
1
46
Z21966.1
-
5
1951
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
heart development
muscle organ development
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0