Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001510.1 | NP_001001510.1 | 3 | 907 | ||
GenBank mRNA | AY495338.1 | - | 3 | 907 | ||
BC085842.1 | - | 3 | 885 | 7135823 |