Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163377.1 | NP_001156849.1 | 3 | 649 | ||
NM_017659.3 | NP_060129.2 | 4 | 931 | |||
XM_017026900.1 | XP_016882389.1 | 4 | 1010 | |||
GenBank mRNA | AK000091.1 | - | 4 | 740 | ||
AK172764.1 | - | 4 | 739 | |||
AK222636.1 | - | 4 | 724 | |||
BC011553.1 | - | 3 | 435 | 4564051 |