Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009676.1 | NP_001009676.1 | 6 | 685 | ||
XM_006245788.1 | XP_006245850.1 | 6 | 728 | |||
XM_006245789.2 | XP_006245851.1 | 6 | 728 | |||
XM_006245790.1 | XP_006245852.1 | 6 | 621 | |||
XM_006245791.1 | XP_006245853.1 | 3 | 332 | |||
XM_008767482.1 | XP_008765704.1 | 6 | 728 | |||
XM_008767483.1 | XP_008765705.1 | 5 | 2457 | |||
XM_008767484.1 | XP_008765706.1 | 4 | 332 | |||
GenBank mRNA | BC087062.1 | - | 6 | 685 | 7131099 |