Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012346.1 | NP_001012346.1 | 6 | 1649 | ||
XM_008771429.1 | XP_008769651.1 | 8 | 2103 | |||
XM_008771430.1 | XP_008769652.1 | 8 | 2103 | |||
XM_008771431.1 | XP_008769653.1 | 7 | 1950 | |||
XM_008771432.1 | XP_008769654.1 | 7 | 1942 | |||
XM_008771433.1 | XP_008769655.1 | 7 | 1815 | |||
GenBank mRNA | AJ810421.1 | - | 6 | 1649 |