Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318052.1 | NP_001304981.1 | 18 | 3390 | ||
NM_015689.4 | NP_056504.3 | 19 | 3492 | |||
NR_134477.1 | 17 | 3059 | ||||
XM_005249977.3 | XP_005250034.1 | 19 | 4023 | |||
XM_011516052.2 | XP_011514354.1 | 19 | 3370 | |||
XM_011516053.2 | XP_011514355.1 | 18 | 3474 | |||
XM_011516054.2 | XP_011514356.1 | 19 | 3275 | |||
XM_017011989.1 | XP_016867478.1 | 18 | 3476 | |||
GenBank mRNA | AB033103.2 | - | 18 | 3390 | ||
AK290056.1 | - | 19 | 3261 | |||
AK297959.1 | - | 11 | 2151 | |||
BC049193.1 | - | 15 | 1860 | |||
BC132875.1 | - | 17 | 3087 | 40146897 |