Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000340.1 | NP_001000340.1 | 1 | 295 |
Gene Symbol: LOC100910858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749523.2 | XP_003749571.1 | 393 |
Gene Symbol: LOC103690075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762014.1 | XP_008760236.1 | 393 |