Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267782.1 | NP_001254711.1 | 2, 3 | 499 | ||
NM_001267783.1 | NP_001254712.1 | 2, 3 | 499 | |||
NM_001300731.1 | NP_001287660.1 | 2, 3 | 499 | |||
NM_017749.3 | NP_060219.2 | 2, 3 | 497 | |||
XM_005253009.3 | XP_005253066.1 | 2, 3 | 495 | |||
XM_005253011.3 | XP_005253068.1 | 2, 3 | 495 | |||
XM_005253014.3 | XP_005253071.1 | 2, 3 | 495 | |||
XM_006718259.2 | XP_006718322.1 | 2, 3 | 376 | |||
XM_006718260.2 | XP_006718323.1 | 2, 3 | 495 | |||
XM_017018007.1 | XP_016873496.1 | 2, 3 | 495 | |||
XM_017018008.1 | XP_016873497.1 | 2, 3 | 495 | |||
GenBank mRNA | AB051523.1 | - | 2, 3 | 451 | ||
AK000301.1 | - | 2, 3 | 450 | |||
AK302808.1 | - | 2, 3 | 557 | |||
AL834190.1 | - | 2, 3 | 378 | |||
BC045609.1 | - | 2, 3 | 497 | 5296472 | ||
DB070055.1 | - | 2, 3 | 499 | |||
DB232750.1 | - | 1, 2 | 42 | |||
DQ870924.1 | - | 1, 2 | 137 |