Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034915.2 | NP_001030087.2 | 9 | 1095 | ||
NM_001122825.1 | NP_001116297.1 | 9 | 1095 | |||
NM_001122826.1 | NP_001116298.1 | 9 | 1095 | |||
NM_001122827.1 | NP_001116299.1 | 9 | 1095 | |||
NM_017697.3 | NP_060167.2 | 9 | 1095 | |||
XM_005250991.3 | XP_005251048.1 | 9 | 1232 | |||
XM_005250992.3 | XP_005251049.1 | 9 | 1232 | |||
GenBank mRNA | AK000178.1 | - | 6 | 489 | ||
BC019932.2 | - | 9 | 1056 | |||
BC035406.1 | - | 6 | 508 | |||
BC067098.1 | - | 9 | 1049 | 6586195 | ||
BC099916.1 | - | 9 | 1059 | 30527936 | ||
BC101621.1 | - | 6 | 454 | |||
BC112043.1 | - | 6 | 454 | |||
BX647570.1 | - | 9 | 1157 |