Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000444.1 | NP_001000444.1 | 637 |
Gene Symbol: LOC100910939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750474.3 | XP_003750522.3 | 1 | 658 |
Gene Symbol: LOC100911888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749656.3 | XP_003749704.3 | 1 | 658 |