siRNA Details
Gene Symbol: Olr1736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001421.1 | NP_001001421.1 | 1 | 737 |

Gene Symbol: Olr1736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001421.1 | NP_001001421.1 | 1 | 737 |